ENA946833930 Enamine

1-(4-bromophenyl)-3-(chloromethyl)-1H-pyrazole

  • MDL number MFCD21304481
  • Cas Number : 1468759-86-5
  • Molecular Weight : 271.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-371142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371142
Availability details for
Quantity
  • Availability
  • Add to Cart