ENA946833945 Enamine

prop-2-en-1-yl methanesulfonate

  • MDL number MFCD01715319
  • Cas Number : 6728-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367064

Safety Information

signalword Danger
pcodes P201,P202,P260,P264,P270,P280,P281,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P308+P313,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-367064
pictograms GHS05,GHS07,GHS08
hstatements H302,H314,H341,H351
Availability details for
Quantity
  • Availability
  • Add to Cart