ENA946833946 Enamine

5-methylfuran-2-carbonyl isothiocyanate

  • MDL number MFCD12188931
  • Cas Number : 856906-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-371330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371330
Availability details for
Quantity
  • Availability
  • Add to Cart