ENA946833950 Enamine

6-bromo-1H,3H,4H,4aH,5H,9bH-thiopyrano[4,3-b]indole

  • MDL number MFCD28537764
  • Cas Number : 1780496-84-5
  • Molecular Weight : 270.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-371675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371675
Availability details for
Quantity
  • Availability
  • Add to Cart