ENA946834010 Enamine

lithium(1+) ion 2,4-dihydroxy-3,3-dimethylbutanoate

  • MDL number MFCD30730679
  • Cas Number : 55289-50-4
  • Molecular Weight : 154.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366956
Availability details for
Quantity
  • Availability
  • Add to Cart