ENA946834832 Enamine

2,5-dihydroxy-1H-pyrrol-1-yl 2-(2,2,3,3,11,11,12,12-octamethyl-4,10-dioxa-7-aza-3,11-disilatridecan-7-yl)acetate

  • MDL number MFCD30731612
  • Cas Number : 2095410-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-373794

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-373794
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart