ENA946834833 Enamine

3-(5-bromo-2-phenyl-1H-indol-3-yl)propanoic acid

  • MDL number MFCD30731613
  • Cas Number : 2094834-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-373937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-373937
Availability details for
Quantity
  • Availability
  • Add to Cart