ENA946834836 Enamine

4H,5H,6H,7H,8H-pyrazolo[1,5-a][1,4]diazepin-2-amine

  • MDL number MFCD30731616
  • Cas Number : 2095409-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374066
Availability details for
Quantity
  • Availability
  • Add to Cart