ENA946834840 Enamine

4-methyl-4-(pyrrolidin-1-yl)cyclohexan-1-one

  • MDL number MFCD30731621
  • Cas Number : 2095410-37-8
  • Molecular Weight : 181.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374397
Availability details for
Quantity
  • Availability
  • Add to Cart