ENA946834841 Enamine

3-(dimethylamino)-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid hydrochloride

  • MDL number MFCD30731622
  • Cas Number : 2094650-63-0
  • Molecular Weight : 257.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374402
Availability details for
Quantity
  • Availability
  • Add to Cart