ENA946834877 Enamine

tert-butyl 4-amino-9-oxa-2-azaspiro[5.5]undecane-2-carboxylate

  • MDL number MFCD30731667
  • Cas Number : 2095410-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374492
Availability details for
Quantity
  • Availability
  • Add to Cart