ENA946835033 Enamine

tert-butyl 3-(4-acetyl-1H-1,2,3-triazol-1-yl)azetidine-1-carboxylate

  • MDL number MFCD30731840
  • Cas Number : 2027580-00-1
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374723
Availability details for
Quantity
  • Availability
  • Add to Cart