ENA946835391 Enamine

2-(chloromethyl)-1,4-dioxaspiro[4.5]decane

  • MDL number MFCD00465683
  • Cas Number : 5503-32-2
  • Molecular Weight : 190.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-373456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-373456
Availability details for
Quantity
  • Availability
  • Add to Cart