ENA946835404 Enamine

2-(quinolin-5-yl)propan-2-ol

  • MDL number MFCD24070915
  • Cas Number : 1523041-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-373792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-373792
Availability details for
Quantity
  • Availability
  • Add to Cart