ENA946835420 Enamine

2-[benzyl(methyl)amino]-1-cyclopropylethan-1-ol

  • MDL number MFCD19606554
  • Cas Number : 1343058-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-374135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-374135
Availability details for
Quantity
  • Availability
  • Add to Cart