ENA946836485 Enamine

3-(oxiran-2-yl)-1-phenylpropan-1-ol

  • MDL number MFCD30732321
  • Cas Number : 2095411-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-375809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-375809
Availability details for
Quantity
  • Availability
  • Add to Cart