ENA946836818 Enamine

3-(5-chloro-2-phenyl-1H-indol-3-yl)propanoic acid

  • MDL number MFCD11589912
  • Cas Number : 290832-30-3
  • Molecular Weight : 299.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-376098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-376098
Availability details for
Quantity
  • Availability
  • Add to Cart