ENA946839778 Enamine

2-{2,9-dioxaspiro[5.5]undecan-3-yl}acetonitrile

  • MDL number MFCD30733998
  • Cas Number : 2095410-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378434
Availability details for
Quantity
  • Availability
  • Add to Cart