ENA946839785 Enamine

(2S,4S)-2-(fluoromethyl)-4-methoxypyrrolidine hydrochloride

  • MDL number MFCD30734007
  • Cas Number : 2095396-35-1
  • Molecular Weight : 169.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378792
Availability details for
Quantity
  • Availability
  • Add to Cart