ENA946839787 Enamine

3-(2-chlorophenyl)-2,2-dimethylpyrrolidine hydrochloride

  • MDL number MFCD30734010
  • Cas Number : 2094419-41-5
  • Molecular Weight : 246.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378851
Availability details for
Quantity
  • Availability
  • Add to Cart