ENA946839805 Enamine

1-nitro-4-(1,2,2,2-tetrafluoroethyl)benzene

  • MDL number MFCD30734041
  • Cas Number : 1637240-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378956
Availability details for
Quantity
  • Availability
  • Add to Cart