ENA946839811 Enamine

tert-butyl N-[(1S,3R,4S)-3,4-dihydroxycyclopentyl]carbamate

  • MDL number MFCD30734049
  • Cas Number : 1854105-57-9
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379060
Availability details for
Quantity
  • Availability
  • Add to Cart