ENA946839814 Enamine

3-(3-phenyl-1H-pyrazol-5-yl)piperidine dihydrochloride

  • MDL number MFCD30734054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379085
Availability details for
Quantity
  • Availability
  • Add to Cart