ENA946839817 Enamine

ethyl 5-(ethylamino)-1,3,4-oxadiazole-2-carboxylate

  • MDL number MFCD30734058
  • Cas Number : 2094590-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379161
Availability details for
Quantity
  • Availability
  • Add to Cart