ENA946839822 Enamine

9-oxa-2-azaspiro[5.5]undecan-4-ol hydrochloride

  • MDL number MFCD30734066
  • Cas Number : 2094607-75-5
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379469
Availability details for
Quantity
  • Availability
  • Add to Cart