ENA946839826 Enamine

3-(bromomethyl)-2,9-dioxaspiro[5.5]undecane

  • MDL number MFCD30734072
  • Cas Number : 2092356-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379493
Availability details for
Quantity
  • Availability
  • Add to Cart