ENA946839834 Enamine

sodium 6-fluoro-2-oxo-1,2,3,4-tetrahydroquinoline-3-carboxylate

  • MDL number MFCD30734088
  • Cas Number : 2094151-61-6
  • Molecular Weight : 231.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379626
Availability details for
Quantity
  • Availability
  • Add to Cart