ENA946839840 Enamine

ethyl 5-[(2-methoxy-2-oxoethyl)sulfanyl]-2,2-dimethylpentanoate

  • MDL number MFCD30734094
  • Cas Number : 2095410-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380817
Availability details for
Quantity
  • Availability
  • Add to Cart