ENA946839841 Enamine

5-(3-methoxyazetidin-3-yl)-1-methyl-1H-1,2,3-triazole dihydrochloride

  • MDL number MFCD30734097
  • Cas Number : 2094860-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380827
Availability details for
Quantity
  • Availability
  • Add to Cart