ENA946839845 Enamine

2-{2-[(1r,4r)-4-methylcyclohexyl]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD30734103
  • Cas Number : 2095396-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380859
Availability details for
Quantity
  • Availability
  • Add to Cart