ENA946839849 Enamine

tert-butyl 7-methyl-6-oxa-2,9-diazaspiro[4.5]decane-2-carboxylate

  • MDL number MFCD30734110
  • Cas Number : 2095409-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-380926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-380926
Availability details for
Quantity
  • Availability
  • Add to Cart