ENA946839867 Enamine

5-fluoro-2-{5-[(piperidin-4-yl)methyl]-1,2,4-oxadiazol-3-yl}pyridine dihydrochloride

  • MDL number MFCD30734132
  • Cas Number : 2095410-64-1
  • Molecular Weight : 335.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381102

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-381102
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart