ENA946839878 Enamine

ethyl 4-methoxy-5-methyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD30734147
  • Cas Number : 2095410-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-381213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-381213
Availability details for
Quantity
  • Availability
  • Add to Cart