ENA946839881 Enamine

4,5-dihydro-3H-spiro[1,4-benzoxazepine-2,1'-cyclobutane]-5-one

  • MDL number MFCD30734150
  • Cas Number : 2094372-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383214
Availability details for
Quantity
  • Availability
  • Add to Cart