ENA946839882 Enamine

1-[(3-cyclopropylpropyl)sulfanyl]ethan-1-one

  • MDL number MFCD30734152
  • Cas Number : 2095410-61-8
  • Molecular Weight : 158.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383261
Availability details for
Quantity
  • Availability
  • Add to Cart