ENA946839887 Enamine

N,N-dimethyl-1,2,3,4-tetrahydro-1,7-naphthyridin-8-amine

  • MDL number MFCD30734158
  • Cas Number : 2095410-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383292
Availability details for
Quantity
  • Availability
  • Add to Cart