ENA946839891 Enamine

methyl (2R)-3-amino-2-phenoxypropanoate hydrochloride

  • MDL number MFCD30734163
  • Cas Number : 2095396-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383757
Availability details for
Quantity
  • Availability
  • Add to Cart