ENA946839892 Enamine

5-(dimethylamino)-2-(hydroxymethyl)-1,3-oxazole-4-carbonitrile

  • MDL number MFCD30734167
  • Cas Number : 2094495-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383844
Availability details for
Quantity
  • Availability
  • Add to Cart