ENA946839894 Enamine

spiro[chromene-2,1'-cyclobutane]-6-amine hydrochloride

  • MDL number MFCD30734169
  • Cas Number : 2094623-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383854
Availability details for
Quantity
  • Availability
  • Add to Cart