ENA946839897 Enamine

(thian-3-yl)hydrazine dihydrochloride

  • MDL number MFCD30734172
  • Cas Number : 2095410-52-7
  • Molecular Weight : 205.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383863
Availability details for
Quantity
  • Availability
  • Add to Cart