ENA946839907 Enamine

(1R,3S,5S)-3-(1-methyl-1H-pyrazol-5-yl)-8-azabicyclo[3.2.1]octan-3-ol

  • MDL number MFCD30734185
  • Cas Number : 1346216-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-383986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-383986
Availability details for
Quantity
  • Availability
  • Add to Cart