ENA946839913 Enamine

methyl 8-amino-5-oxaspiro[3.5]nonane-2-carboxylate

  • MDL number MFCD30734192
  • Cas Number : 2095409-14-4
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384051
Availability details for
Quantity
  • Availability
  • Add to Cart