ENA946839917 Enamine

3-iodo-2-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD30734196
  • Cas Number : 2095408-90-3
  • Molecular Weight : 263.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384066
Availability details for
Quantity
  • Availability
  • Add to Cart