ENA946839925 Enamine

2-[(pyridin-2-yl)methoxy]acetic acid; trifluoroacetic acid

  • MDL number MFCD30734207
  • Cas Number : 2095409-44-0
  • Molecular Weight : 281.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384170
Availability details for
Quantity
  • Availability
  • Add to Cart