ENA946839929 Enamine

3-(1H-1,2,3-triazol-5-yl)azetidin-3-ol dihydrochloride

  • MDL number MFCD30734214
  • Cas Number : 2094164-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384234
Availability details for
Quantity
  • Availability
  • Add to Cart