ENA946839947 Enamine

2-fluoro-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD24520191
  • Cas Number : 1824337-84-9
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378879
Availability details for
Quantity
  • Availability
  • Add to Cart