ENA946839961 Enamine

N-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)-4-(methylamino)-3-nitrobenzene-1-sulfonamide

  • MDL number MFCD30734025
  • Cas Number : 1512443-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378907
Availability details for
Quantity
  • Availability
  • Add to Cart