ENA946839972 Enamine

tert-butyl (2S,4S)-4-fluoro-2-[(methanesulfonyloxy)methyl]pyrrolidine-1-carboxylate

  • MDL number MFCD30734033
  • Cas Number : 1033245-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378926
Availability details for
Quantity
  • Availability
  • Add to Cart