ENA946839996 Enamine

methyl 4-oxo-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number MFCD26382166
  • Cas Number : 1708268-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-378981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378981
Availability details for
Quantity
  • Availability
  • Add to Cart