ENA946840010 Enamine

2-(1-benzofuran-7-yl)acetonitrile

  • MDL number MFCD15146000
  • Cas Number : 84549-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-379042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-379042
Availability details for
Quantity
  • Availability
  • Add to Cart